Learn More
Biomatik Corporation Human Defensin Alpha 3, Neutrophil Specific (DEFa3) ELISA Kit, 96 tests
Catalog No. 501502316
Encompass_Preferred
The Human Defensin Alpha 3, Neutrophil Specific (DEFa3) ELISA Kit allows for quantitative determination of Defensin Alpha 3, Neutrophil Specific in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.47-30 ng/mL. Sensitivity: 0.21 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P59666. Cross Activity: No significant cross-reactivity or interference between Defensin Alpha 3, Neutrophil Specific and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2316 Supplier Biomatik Corporation Supplier No. EKU03650
The Human Defensin Alpha 3, Neutrophil Specific (DEFa3) ELISA Kit allows for quantitative determination of Defensin Alpha 3, Neutrophil Specific in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.47-30 ng/mL. Sensitivity: 0.21 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P59666. Cross Activity: No significant cross-reactivity or interference between Defensin Alpha 3, Neutrophil Specific and analogues was observed. Restriction: For Research Use Only.