Learn More
Biomatik Corporation Cattle Defensin Beta 2 (DEFb2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03667
Encompass_Preferred
The Cattle Defensin Beta 2 (DEFb2) ELISA Kit allows for quantitative determination of Defensin Beta 2 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: core/N. Cross Activity: No significant cross-reactivity or interference between Defensin Beta 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2321