Learn More
Biomatik Corporation Human Delta Like 1 Homolog (dLK1) ELISA Kit, 96 tests
Catalog No. 501502326
Encompass_Preferred
The Human Delta Like 1 Homolog (dLK1) ELISA Kit allows for quantitative determination of Delta Like 1 Homolog in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.243 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P80370. Cross Activity: No significant cross-reactivity or interference between Delta Like 1 Homolog and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2326 Supplier Biomatik Corporation Supplier No. EKU03680
The Human Delta Like 1 Homolog (dLK1) ELISA Kit allows for quantitative determination of Delta Like 1 Homolog in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.243 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P80370. Cross Activity: No significant cross-reactivity or interference between Delta Like 1 Homolog and analogues was observed. Restriction: For Research Use Only.