Learn More
Biomatik Corporation Rat Delta Like 1 Homolog (dLK1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03682
Encompass_Preferred
Rat Delta Like 1 Homolog (dLK1) ELISA Kit for detecting dLK1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.27ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O70534. Gene ID: 114587. Target Synonyms: FA1; PREF1; Pref-1; ZOG; pG2; Protein delta homolog 1; Fetal antigen 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2328