Learn More
Biomatik Corporation Cattle Deoxyribonuclease I (DNASE1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03690
Encompass_Preferred
The Cattle Deoxyribonuclease I (DNASE1) ELISA Kit allows for quantitative determination of Deoxyribonuclease I in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.62 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00639. Cross Activity: No significant cross-reactivity or interference between Deoxyribonuclease I and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2329