Learn More
Biomatik Corporation Human Deoxyribonuclease I (DNASE1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03691
Encompass_Preferred
The Human Deoxyribonuclease I (DNASE1) ELISA Kit allows for quantitative determination of Deoxyribonuclease I in Serum, Plasma, Urine, Saliva, seminal Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 78-5,000 pg/mL. Sensitivity: 30 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P24855. Cross Activity: No significant cross-reactivity or interference between Deoxyribonuclease I and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2330