Learn More
Biomatik Corporation Mouse Deoxyribonuclease I (DNASE1) ELISA Kit, 96 tests
Catalog No. 501502331
Encompass_Preferred
Mouse Deoxyribonuclease I (DNASE1) ELISA Kit for detecting DNASE1 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.25-80ng/ml. Sensitivity: 0.45ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49183. Gene ID: 13419. Target Synonyms: DNaseI; DNL1; DRNI; DNase-I; Dornase alfa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2331 Supplier Biomatik Corporation Supplier No. EKU03692
Mouse Deoxyribonuclease I (DNASE1) ELISA Kit for detecting DNASE1 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.25-80ng/ml. Sensitivity: 0.45ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49183. Gene ID: 13419. Target Synonyms: DNaseI; DNL1; DRNI; DNase-I; Dornase alfa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.