Learn More
Biomatik Corporation Pig Diamine Oxidase (DAO) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03721
Encompass_Preferred
Pig Amiloride Binding Protein 1 (ABP1) ELISA Kit for detecting ABP1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 3.12-200ng/ml. Sensitivity: 1.17ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9TRC7. Gene ID: 100517436. Target Synonyms: AOC1; DAO; KAO; Histaminase; Kidney amine oxidase; Diamine Oxidase; Amiloride-sensitive amine oxidase; Amine oxidase copper domain-containing protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2344