Learn More
Biomatik Corporation Mouse Diazepam Binding Inhibitor (DBI) ELISA Kit, 96 tests
Catalog No. 501502347
Encompass_Preferred
Mouse Diazepam Binding Inhibitor (DBI) ELISA Kit for detecting DBI in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500ng/ml. Sensitivity: 3.1ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31786. Gene ID: 13167. Target Synonyms: ACBP; ACBD1; CCK-RP; EP; Endozepine; GABA Receptor Modulator; Acyl-Coenzyme A Binding Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2347 Supplier Biomatik Corporation Supplier No. EKU03725
Mouse Diazepam Binding Inhibitor (DBI) ELISA Kit for detecting DBI in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500ng/ml. Sensitivity: 3.1ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31786. Gene ID: 13167. Target Synonyms: ACBP; ACBD1; CCK-RP; EP; Endozepine; GABA Receptor Modulator; Acyl-Coenzyme A Binding Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.