Learn More
Biomatik Corporation Human Dicer 1, Ribonuclease Type III (DICER1) ELISA Kit, 96 tests
Catalog No. 501502348
Encompass_Preferred
The Human Dicer 1, Ribonuclease Type III (DICER1) ELISA Kit allows for quantitative determination of Dicer 1, Ribonuclease Type III in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.112 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9UPY3. Cross Activity: No significant cross-reactivity or interference between Dicer 1, Ribonuclease Type III and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2348 Supplier Biomatik Corporation Supplier No. EKU03728
The Human Dicer 1, Ribonuclease Type III (DICER1) ELISA Kit allows for quantitative determination of Dicer 1, Ribonuclease Type III in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.112 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9UPY3. Cross Activity: No significant cross-reactivity or interference between Dicer 1, Ribonuclease Type III and analogues was observed. Restriction: For Research Use Only.