Learn More
Biomatik Corporation Human Dihydropyrimidine Dehydrogenase (DPYD) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03741
Encompass_Preferred
The Human Dihydropyrimidine Dehydrogenase (DPYD) ELISA Kit allows for quantitative determination of Dihydropyrimidine Dehydrogenase in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-1000 pg/mL. Sensitivity: 26.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q12882. Cross Activity: No significant cross-reactivity or interference between Dihydropyrimidine Dehydrogenase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2352