Learn More
Biomatik Corporation Human Dipeptidyl Peptidase IV (DPP4) ELISA Kit, 96 tests
Catalog No. 501502355
Encompass_Preferred
The Human Dipeptidyl Peptidase IV (DPP4) ELISA Kit allows for quantitative determination of Dipeptidyl Peptidase IV in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.253 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P27487. Cross Activity: No significant cross-reactivity or interference between Dipeptidyl Peptidase IV and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2355 Supplier Biomatik Corporation Supplier No. EKU03755
The Human Dipeptidyl Peptidase IV (DPP4) ELISA Kit allows for quantitative determination of Dipeptidyl Peptidase IV in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.253 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P27487. Cross Activity: No significant cross-reactivity or interference between Dipeptidyl Peptidase IV and analogues was observed. Restriction: For Research Use Only.