Learn More
Biomatik Corporation Rat Dipeptidyl Peptidase IV (DPP4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03757
Encompass_Preferred
Rat Cluster Of Differentiation 26 (CD26) ELISA Kit for detecting CD26 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.269ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P14740. Gene ID: 25253. Target Synonyms: CD26; ADA; DPPIV; DPP-IV; DPP4; ADABP; ADCP2; TP103; Adenosine Deaminase Complexing Protein 2; T-cell activation antigen CD26; Dipeptidyl Peptidase IV. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2356