Learn More
Biomatik Corporation Mouse Discs, Large Homolog 4 (DLG4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03761
Encompass_Preferred
Mouse Discs, Large Homolog 4 (DLG4) ELISA Kit for detecting DLG4 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62108. Gene ID: 13385. Target Synonyms: PSD95; SAP90; Postsynaptic density protein 95; Synapse-associated protein 90. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2358