Learn More
Biomatik Corporation Human DNA Damage Inducible Transcript 3 (DDIT3) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU03766
The Human DNA Damage Inducible Transcript 3 (DDIT3) ELISA Kit allows for quantitative determination of DNA Damage Inducible Transcript 3 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.112 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P35638. Cross Activity: No significant cross-reactivity or interference between DNA Damage Inducible Transcript 3 and analogues was observed. Restriction: For Research Use Only.