Learn More
Biomatik Corporation Mouse DNA Damage Inducible Transcript 3 (DDIT3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03767
Encompass_Preferred
Mouse DNA Damage Inducible Transcript 3 (DDIT3) ELISA Kit for detecting DDIT3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35639. Gene ID: 13198. Target Synonyms: CEBPZ; CHOP; CHOP10; GADD153; C/EBP-homologous protein 10; CCAAT/enhancer-binding protein homologous protein; Growth arrest and DNA damage-inducible GADD153. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2360