Learn More
Biomatik Corporation Human Dopamine Receptor D2 (DRD2) ELISA Kit, 96 tests
Catalog No. 501502364
Encompass_Preferred
The Human Dopamine Receptor D2 (DRD2) ELISA Kit allows for quantitative determination of Dopamine Receptor D2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.123 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P14416. Cross Activity: No significant cross-reactivity or interference between Dopamine Receptor D2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2364 Supplier Biomatik Corporation Supplier No. EKU03783
The Human Dopamine Receptor D2 (DRD2) ELISA Kit allows for quantitative determination of Dopamine Receptor D2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.123 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P14416. Cross Activity: No significant cross-reactivity or interference between Dopamine Receptor D2 and analogues was observed. Restriction: For Research Use Only.