Learn More
Biomatik Corporation Rat Dopamine Receptor D4 (DRD4) ELISA Kit, 96 tests
Catalog No. 501502365
Encompass_Preferred
Rat Dopamine Receptor D4 (DRD4) ELISA Kit for detecting DRD4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P30729. Target Synonyms: DR-D4; D4R; D4DR; D (2C) dopamine receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2365 Supplier Biomatik Corporation Supplier No. EKU03786
Rat Dopamine Receptor D4 (DRD4) ELISA Kit for detecting DRD4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P30729. Target Synonyms: DR-D4; D4R; D4DR; D (2C) dopamine receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.