Learn More
Biomatik Corporation Human Dual Specificity Phosphatase 1 (DUSP1) ELISA Kit, 96 tests
Catalog No. 501502369
Encompass_Preferred
The Human Dual Specificity Phosphatase 1 (DUSP1) ELISA Kit allows for quantitative determination of Dual Specificity Phosphatase 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.25-2000 pg/mL. Sensitivity: 12.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P28562. Cross Activity: No significant cross-reactivity or interference between Dual Specificity Phosphatase 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2369 Supplier Biomatik Corporation Supplier No. EKU03798
The Human Dual Specificity Phosphatase 1 (DUSP1) ELISA Kit allows for quantitative determination of Dual Specificity Phosphatase 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.25-2000 pg/mL. Sensitivity: 12.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P28562. Cross Activity: No significant cross-reactivity or interference between Dual Specificity Phosphatase 1 and analogues was observed. Restriction: For Research Use Only.