Learn More
Biomatik Corporation Mouse E1A Binding Protein P300 (EP300) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03819
Encompass_Preferred
Mouse E1A Binding Protein P300 (EP300) ELISA Kit for detecting EP300 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: B2RWS6. Gene ID: 328572. Target Synonyms: KAT3B; p300; Histone Acetyltransferase p300. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2374