Learn More
Biomatik Corporation Human EGF Like Domain Protein, Multiple 6 (EGFL6) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03831
Encompass_Preferred
The Human EGF Like Domain Protein, Multiple 6 (EGFL6) ELISA Kit allows for quantitative determination of EGF Like Domain Protein, Multiple 6 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 93.75-6000 pg/mL. Sensitivity: 35.95 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8IUX8. Cross Activity: No significant cross-reactivity or interference between EGF Like Domain Protein, Multiple 6 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2375