Learn More
Biomatik Corporation Mouse EGF Like Domain Protein, Multiple 7 (EGFL7) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03833
Encompass_Preferred
Mouse EGF Like Domain Protein, Multiple 7 (EGFL7) ELISA Kit for detecting EGFL7 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QXT5. Gene ID: 353156. Target Synonyms: ZNEU1; MEGF7; Zneu1; Vascular endothelial statin; EGF Like Domain Containing 7; Epidermal growth factor-like protein 7; Multiple epidermal growth factor-like domains protein 7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2377