Learn More
Biomatik Corporation Human EGF Like Repeats And Discoidin I Like Domains Protein 3 (EDIL3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03835
Encompass_Preferred
The Human EGF Like Repeats And Discoidin I Like Domains Protein 3 (EDIL3) ELISA Kit allows for quantitative determination of EGF Like Repeats And Discoidin I Like Domains Protein 3 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.127 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O43854. Cross Activity: No significant cross-reactivity or interference between EGF Like Repeats And Discoidin I Like Domains Protein 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2379