Learn More
Biomatik Corporation Dog Elastase 2, Neutrophil (ELA2) ELISA Kit, 96 tests
Catalog No. 501502380
Encompass_Preferred
The Dog Elastase 2, Neutrophil (ELA2) ELISA Kit allows for quantitative determination of Elastase 2, Neutrophil in Serum, Plasma and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 31.2-2000 pg/mL. Sensitivity: 12.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8MJD1. Cross Activity: No significant cross-reactivity or interference between Elastase 2, Neutrophil and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2380 Supplier Biomatik Corporation Supplier No. EKU03840
The Dog Elastase 2, Neutrophil (ELA2) ELISA Kit allows for quantitative determination of Elastase 2, Neutrophil in Serum, Plasma and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 31.2-2000 pg/mL. Sensitivity: 12.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8MJD1. Cross Activity: No significant cross-reactivity or interference between Elastase 2, Neutrophil and analogues was observed. Restriction: For Research Use Only.