Learn More
Biomatik Corporation Human Elastase 2, Neutrophil (ELA2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03841
Encompass_Preferred
The Human Elastase 2, Neutrophil (ELA2) ELISA Kit allows for quantitative determination of Elastase 2, Neutrophil in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.19-12 ng/mL. Sensitivity: 0.07 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08246. Cross Activity: No significant cross-reactivity or interference between Elastase 2, Neutrophil and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2381