Learn More
Biomatik Corporation Mouse Elastase 2, Neutrophil (ELA2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03842
Encompass_Preferred
Mouse Neutrophil Elastase (NE) ELISA Kit for detecting NE in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q3UP87. Gene ID: 50701. Target Synonyms: ELANE; HLE; HNE; ELA2; PMN-E; Neutrophil Elastase; Medullasin; Polymorphonuclear Leukocyte Elastase; Bone Marrow Serine Protease; Human Leukocyte Elastase; PMN Elastase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2382