Learn More
Biomatik Corporation Rat Elastase 2, Neutrophil (ELA2) ELISA Kit, 96 tests
Catalog No. 501502383
Encompass_Preferred
Rat Neutrophil Elastase (NE) ELISA Kit for detecting NE in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500ng/ml. Sensitivity: 3.3ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D4A488. Gene ID: 299606. Target Synonyms: ELANE; HLE; HNE; ELA2; PMN-E; Neutrophil Elastase; Medullasin; Polymorphonuclear Leukocyte Elastase; Bone Marrow Serine Protease; Human Leukocyte Elastase; PMN Elastase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2383 Supplier Biomatik Corporation Supplier No. EKU03843
Rat Neutrophil Elastase (NE) ELISA Kit for detecting NE in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500ng/ml. Sensitivity: 3.3ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D4A488. Gene ID: 299606. Target Synonyms: ELANE; HLE; HNE; ELA2; PMN-E; Neutrophil Elastase; Medullasin; Polymorphonuclear Leukocyte Elastase; Bone Marrow Serine Protease; Human Leukocyte Elastase; PMN Elastase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.