Learn More
Biomatik Corporation Human Endoglin (ENG) ELISA Kit, 96 tests
Catalog No. 501502390
Encompass_Preferred
The Human Endoglin (ENG) ELISA Kit allows for quantitative determination of Endoglin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.063 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P17813. Cross Activity: No significant cross-reactivity or interference between Endoglin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2390 Supplier Biomatik Corporation Supplier No. EKU03872
The Human Endoglin (ENG) ELISA Kit allows for quantitative determination of Endoglin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.063 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P17813. Cross Activity: No significant cross-reactivity or interference between Endoglin and analogues was observed. Restriction: For Research Use Only.