Learn More
Biomatik Corporation Human Endothelial Cell Specific Molecule 1 (ESM1) ELISA Kit, 96 tests
Catalog No. 501502391
Encompass_Preferred
The Human Endothelial Cell Specific Molecule 1 (ESM1) ELISA Kit allows for quantitative determination of Endothelial Cell Specific Molecule 1 in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9NQ30. Cross Activity: No significant cross-reactivity or interference between Endothelial Cell Specific Molecule 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2391 Supplier Biomatik Corporation Supplier No. EKU03883
The Human Endothelial Cell Specific Molecule 1 (ESM1) ELISA Kit allows for quantitative determination of Endothelial Cell Specific Molecule 1 in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9NQ30. Cross Activity: No significant cross-reactivity or interference between Endothelial Cell Specific Molecule 1 and analogues was observed. Restriction: For Research Use Only.