Learn More
Biomatik Corporation Human Endothelin 1 (EDN1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03887
Encompass_Preferred
The Human Endothelin 1 (EDN1) ELISA Kit allows for quantitative determination of Endothelin 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 6.17-500 pg/mL. Sensitivity: 2.71 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P05305. Cross Activity: No significant cross-reactivity or interference between Endothelin 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2392