Learn More
Biomatik Corporation Rabbit Endothelin 1 (EDN1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03890
Encompass_Preferred
Rabbit Endothelin 1 (EDN1) ELISA Kit for detecting EDN1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 6.17-500pg/ml. Sensitivity: 2.71pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P29560. Gene ID: 100009270. Target Synonyms: ET1; PPET1; Preproendothelin-1; Big endothelin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2395