Learn More
Biomatik Corporation Human Endothelin Converting Enzyme 1 (ECE1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03892
Encompass_Preferred
The Human Endothelin Converting Enzyme 1 (ECE1) ELISA Kit allows for quantitative determination of Endothelin Converting Enzyme 1 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.25-2000 pg/mL. Sensitivity: 11.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P42892. Cross Activity: No significant cross-reactivity or interference between Endothelin Converting Enzyme 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2396