Learn More
Biomatik Corporation Rat Enolase, Neuron Specific (NSE) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03905
Encompass_Preferred
Rat Enolase, Neuron Specific (NSE) ELISA Kit for detecting NSE in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.253ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07323. Gene ID: 24334. Target Synonyms: ENO2; Enolase 2; Gamma Enolase; 2-phospho-D-glycerate hydro-lyase; Neural enolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2399