Learn More
Biomatik Corporation Human Eosinophil Chemotactic Factor (ECF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03914
Encompass_Preferred
The Human Eosinophil Chemotactic Factor (ECF) ELISA Kit allows for quantitative determination of Eosinophil Chemotactic Factor in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q6I9T4. Cross Activity: No significant cross-reactivity or interference between Eosinophil Chemotactic Factor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2401