Learn More
Biomatik Corporation Mouse Eosinophil Peroxidase (EPX) ELISA Kit, 96 tests
Catalog No. 501502404
Encompass_Preferred
Mouse Eosinophil Peroxidase (EPX) ELISA Kit for detecting EPX in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49290. Gene ID: 13861. Target Synonyms: EPO; EPP; EPX-PEN; EPER. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2404 Supplier Biomatik Corporation Supplier No. EKU03919
Mouse Eosinophil Peroxidase (EPX) ELISA Kit for detecting EPX in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49290. Gene ID: 13861. Target Synonyms: EPO; EPP; EPX-PEN; EPER. Restrictions: For Research Use Only. Not for use in diagnostic procedures.