Learn More
Biomatik Corporation Human Ephrin B2 (EFNB2) ELISA Kit, 96 tests
Catalog No. 501502406
Encompass_Preferred
The Human Ephrin B2 (EFNB2) ELISA Kit allows for quantitative determination of Ephrin B2 in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.062 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P52799. Cross Activity: No significant cross-reactivity or interference between Ephrin B2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2406 Supplier Biomatik Corporation Supplier No. EKU03925
The Human Ephrin B2 (EFNB2) ELISA Kit allows for quantitative determination of Ephrin B2 in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.062 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P52799. Cross Activity: No significant cross-reactivity or interference between Ephrin B2 and analogues was observed. Restriction: For Research Use Only.