Learn More
Biomatik Corporation Human Epidermal Growth Factor (EGF) ELISA Kit, 96 tests
Catalog No. 501502407
Encompass_Preferred
The Human Epidermal Growth Factor (EGF) ELISA Kit allows for quantitative determination of Epidermal Growth Factor in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.9-250 pg/mL. Sensitivity: 1.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01133. Cross Activity: No significant cross-reactivity or interference between Epidermal Growth Factor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2407 Supplier Biomatik Corporation Supplier No. EKU03932
The Human Epidermal Growth Factor (EGF) ELISA Kit allows for quantitative determination of Epidermal Growth Factor in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.9-250 pg/mL. Sensitivity: 1.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01133. Cross Activity: No significant cross-reactivity or interference between Epidermal Growth Factor and analogues was observed. Restriction: For Research Use Only.