Learn More
Biomatik Corporation Mouse Epidermal Growth Factor (EGF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03933
Encompass_Preferred
Mouse Epidermal Growth Factor (EGF) ELISA Kit for detecting EGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 2.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01132. Gene ID: 13645. Target Synonyms: URG; Beta-Urogastrone; Pro-epidermal growth factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2408