Learn More
Biomatik Corporation Rat Epidermal Growth Factor (EGF) ELISA Kit, 96 tests
Catalog No. 501502410
Encompass_Preferred
Rat Epidermal Growth Factor (EGF) ELISA Kit for detecting EGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07522. Gene ID: 25313. Target Synonyms: URG; Beta-Urogastrone; Pro-epidermal growth factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2410 Supplier Biomatik Corporation Supplier No. EKU03935
Rat Epidermal Growth Factor (EGF) ELISA Kit for detecting EGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07522. Gene ID: 25313. Target Synonyms: URG; Beta-Urogastrone; Pro-epidermal growth factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.