Learn More
Biomatik Corporation Human Epidermal Growth Factor Receptor (EGFR) ELISA Kit, 96 tests
Catalog No. 501502411
Encompass_Preferred
The Human Epidermal Growth Factor Receptor (EGFR) ELISA Kit allows for quantitative determination of Epidermal Growth Factor Receptor in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.17 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00533. Cross Activity: No significant cross-reactivity or interference between Epidermal Growth Factor Receptor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2411 Supplier Biomatik Corporation Supplier No. EKU03936
The Human Epidermal Growth Factor Receptor (EGFR) ELISA Kit allows for quantitative determination of Epidermal Growth Factor Receptor in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.17 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00533. Cross Activity: No significant cross-reactivity or interference between Epidermal Growth Factor Receptor and analogues was observed. Restriction: For Research Use Only.