Learn More
Biomatik Corporation Human Epidermal Growth Factor Receptor 2 (EGFR2) ELISA Kit, 96 tests
Catalog No. 501502412
Encompass_Preferred
The Human Epidermal Growth Factor Receptor 2 (EGFR2) ELISA Kit allows for quantitative determination of Epidermal Growth Factor Receptor 2 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.229 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P04626. Cross Activity: No significant cross-reactivity or interference between Epidermal Growth Factor Receptor 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2412 Supplier Biomatik Corporation Supplier No. EKU03939
The Human Epidermal Growth Factor Receptor 2 (EGFR2) ELISA Kit allows for quantitative determination of Epidermal Growth Factor Receptor 2 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.229 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P04626. Cross Activity: No significant cross-reactivity or interference between Epidermal Growth Factor Receptor 2 and analogues was observed. Restriction: For Research Use Only.