Learn More
Biomatik Corporation Epinephrine (EPI) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03943
Encompass_Preferred
The Epinephrine (EPI) ELISA Kit allows for quantitative determination of Epinephrine in Serum, Plasma, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: All species. Detection Range: 24.69-2000 pg/mL. Sensitivity: 9.74 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Epinephrine and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2413