Learn More
Biomatik Corporation Mouse Androgen Receptor (AR) ELISA Kit
Supplier:  Biomatik Corporation EKU02343
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P19091. Target Synonyms: AIS; DHTR; HUMARA; KD; NR3C4; SBMA; SMAX1; TFM; Dihydrotestosterone receptor; Nuclear Receptor Subfamily 3,Group C,Member 4; Testicular Feminization; Kennedy Disease. Restriction: For Research Use Only.
Catalog No. 50-228-5791