Learn More
Biomatik Corporation Human Arylsulfatase A (ARSA) ELISA Kit
Supplier:  Biomatik Corporation EKU02583
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P15289. Target Synonyms: MLD; ASA; Metachromatic Leucodystrophy; Cerebroside-sulfatase. Restriction: For Research Use Only.
Catalog No. 50-228-5821