Learn More
Biomatik Corporation Dog Erythropoietin (EPO) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03960
Encompass_Preferred
The Dog Erythropoietin (EPO) ELISA Kit allows for quantitative determination of Erythropoietin in Serum, Plasma and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 31.2-2000 pg/mL. Sensitivity: 13.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P33707. Cross Activity: No significant cross-reactivity or interference between Erythropoietin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2416