Learn More
Biomatik Corporation Mouse Cereblon (CRBN) ELISA Kit
Supplier:  Biomatik Corporation EKU03078
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q8C7D2. Target Synonyms: MRT2A; Mental Retardation,Non-Syndromic,Autosomal Recessive 2A. Restriction: For Research Use Only.
Catalog No. 50-228-5876