Learn More
Biomatik Corporation Human Erythropoietin (EPO) ELISA Kit, 96 tests
Catalog No. 501502418
Encompass_Preferred
The Human Erythropoietin (EPO) ELISA Kit allows for quantitative determination of Erythropoietin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2,000 pg/mL. Sensitivity: 11.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01588. Cross Activity: No significant cross-reactivity or interference between Erythropoietin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2418 Supplier Biomatik Corporation Supplier No. EKU03962
The Human Erythropoietin (EPO) ELISA Kit allows for quantitative determination of Erythropoietin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2,000 pg/mL. Sensitivity: 11.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01588. Cross Activity: No significant cross-reactivity or interference between Erythropoietin and analogues was observed. Restriction: For Research Use Only.