Learn More
Biomatik Corporation Mouse Cluster Of Differentiation 14 (CD14) ELISA Kit
Supplier:  Biomatik Corporation EKU03224
Encompass_Preferred
Mouse Cluster Of Differentiation 14 (CD14) ELISA Kit for detecting CD14 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 29.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10810. Gene ID: 12475. Target Synonyms: Monocyte differentiation antigen CD14; Myeloid cell-specific leucine-rich glycoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5894