Learn More
Biomatik Corporation Rat Erythropoietin (EPO) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03965
Encompass_Preferred
Rat Erythropoietin (EPO) ELISA Kit for detecting EPO in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 93.75-6,000pg/ml. Sensitivity: 33.59pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29676. Gene ID: 24335. Target Synonyms: EP; Epoetin; Erythropoetin; Hematopoietin; Hemopoietin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2421