Learn More
Biomatik Corporation Mouse Collagen Type IV (COL4) ELISA Kit
Supplier:  Biomatik Corporation EKU03322
Encompass_Preferred
Mouse Collagen Type IV (COL4) ELISA Kit for detecting COL4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02463; P08122. Gene ID: 12826; 12827. Target Synonyms: Type-IV Collagen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5906